Biopsy processing/embedding cassettes, Slimsette®, M510
Gul
Catalog # 720-1608
Supplier: Simport Scientific
Biopsy processing/embedding cassettes, Slimsette®, M510
Catalog # 720-1608
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Acetal polymer. Cassette design is similar to that of the M509 model but specially designed to hold biopsy specimens.
- Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
- Disposable plastic cassettes are highly resistant to histological solutions
- Small, square openings for good fluid exchange
- Integral, recessed lid
- Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
Packaging: Each case contains three boxes holding 500 cassettes each.
Specifications
- Färg:Gul