Vacuum stopcocks
Vacuum stopcocks, one way, right-angled, 4,0 mm, 18,8 NS
Katalog # 189-2122
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
- Excellent chemical resistance
- Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Hål Ø.:4,0 mm
- Beskrivning:Vacuum stopcocks, one way, right-angled
- Seq:12
- Kran:18,8 NS