Orders LinkContactUsLinkComponent
Solid phase microextraction fibers
Solid phase microextraction fibers
SPME fiber for manual use, carboxen/PDMS, 24 gauge, 75 µm fiber coating
Katalog # VARI391896315
Leverantör:   VARIAN
Solid phase microextraction fibers
Katalog # VARI391896315
Leverantör:  VARIAN
Solid phase microextraction fibers
SPME fiber for manual use, carboxen/PDMS, 24 gauge, 75 µm fiber coating
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 3 Items
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifikationer
  • Beskrivning:
    SPME fiber for manual use, carboxen/PDMS, 24 gauge, 75 µm fiber coating
  • Seq:
    09
Accessories
Produktinformation
  • Katalog #
    Tillgänglighet
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 10 511,32 kr
Produktinformation
  • Katalog #
    Tillgänglighet
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items
Produktinformation
  • Katalog #
    Tillgänglighet
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items
Produktinformation
  • Katalog #
    Tillgänglighet
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U