Orders LinkContactUsLinkComponent
Separating funnels, pear shaped
Separating funnels, pear shaped
2000 ml, 34/35 NS
Katalog # 511-0758
Leverantör:   SciLabware
Separating funnels, pear shaped
Katalog # 511-0758
Leverantör:  SciLabware
Separating funnels, pear shaped
Separating funnels, pear shaped
2000 ml, 34/35 NS
Price
2 625,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Very good chemical resistance
  • Very good temperature resistance
Available with ground glass stopper and interchangeable PTFE key.
Specifikationer
  • NS-hylsa:
    34/35 NS
  • Kapacitet:
    2000 ml