Calibration weights, cylindrical, OIML, class E2
100 g, 0,16 ± mg, Cylindrical test weight
Katalog # SARBYCW5128-02
Leverantör: Sartorius Balances
Calibration weights, cylindrical, OIML, class E2
Katalog # SARBYCW5128-02
Leverantör: Sartorius Balances
Gå till:
Produktdetaljer och dokument
Class E2 cyclindrical calibration weights.
Specifikationer
- Vikt:100 g
- Seq:16
- Tolerans:0,16 ± mg
- Typ:Cylindrical test weight