Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall
75 mm, 12 mm
Katalog # 212-7088
Leverantör:   MURRAY S
Test tubes without rim, medium wall
Katalog # 212-7088
Leverantör:  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall
75 mm, 12 mm
Price
570,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Produktdetaljer och dokument
Soda-lime glass, rimless.
  • Good chemical and physical properties
  • Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Specifikationer
  • Ø utv.:
    12 mm
  • Längd:
    75 mm
  • Seq:
    05