Test tubes without rim, medium wall
75 mm, 16 mm
Katalog # 212-7094
Leverantör: MURRAY S
Gå till:
Produktdetaljer och dokument
Soda-lime glass, rimless.
- Good chemical and physical properties
- Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Specifikationer
- Ø utv.:16 mm
- Längd:75 mm
- Seq:06