Test tubes without rim, medium wall
150 mm, 24 mm
Katalog # 212-7093
Leverantör: MURRAY S
Gå till:
Produktdetaljer och dokument
Soda-lime glass, rimless.
- Good chemical and physical properties
- Medium-walled
Suitable for use in applications that do not involve high thermal stresses, especially for bacteriological applications.
Specifikationer
- Ø utv.:24 mm
- Längd:150 mm
- Seq:14