Orders LinkContactUsLinkComponent
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
65 mm, 3, 19/26 NS
Katalog # 201-8681
Leverantör:   SciLabware
Büchner funnels, Quickfit®
Katalog # 201-8681
Leverantör:  SciLabware
Büchner funnels, Quickfit®
Büchner funnels, Quickfit®
65 mm, 3, 19/26 NS
Price
1 250,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With sintered glass disc and integral ground glass cone.
Specifikationer
  • Kärna:
    19/26 NS
  • Platt Ø:
    65 mm
  • Porositet:
    3