Orders LinkContactUsLinkComponent
Bone cutters
Bone cutters
With overlapping spring, 140 mm
Katalog # 233-0010
Leverantör:   MURRAY S
Bone cutters
Katalog # 233-0010
Leverantör:  MURRAY S
Bone cutters
Bone cutters
With overlapping spring, 140 mm
Price
1 150,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Carbon steel, nickel plated. With overlapping spring or coil spring handle.
  • Excellent thermal resistance
  • Easy to clean
  • Safe, simple operation
Specifikationer
  • Beskrivning:
    With overlapping spring
  • Längd:
    140 mm
  • Seq:
    01