Orders LinkContactUsLinkComponent
Column packing support, Quickfit®
Column packing support, Quickfit®
21 mm, 24/29 NS
Katalog # 201-8369
Leverantör:   SciLabware
Column packing support, Quickfit®
Katalog # 201-8369
Leverantör:  SciLabware
Column packing support, Quickfit®
Column packing support, Quickfit®
21 mm, 24/29 NS
Price
250,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
Specifikationer
  • Kärna:
    24/29 NS
  • Ø:
    21 mm
  • Seq:
    02