Soxhlet extractors, complete assemblies
600 ml, 2000 ml, -* NS
Catalog # BIBB600RDSX
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Complete glass assembly
- Low solvent volumes required for extraction of soluble components in solids
- Range of sizes of extractor to meet different sample sizes
* 75 mm diameter extractor flange.
Delivery information: Assembly consists of round bottom flask, Soxhlet extractor and Allihn condenser.
Specifications
- Extraktionsvolym:600 ml
- NS-hylsa:-* NS
- Seq:06
- Volym:2000 ml