Urea
Glass bottle for solids, 250 g
AMBI9900
Marketsource
: Ambion
: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- :57-13-6
- :NH₂CONH₂
- :MFCD00008022
- :60.06 g/mol
- :13,09935
- :197 °C (1013 hPa)
- :132,7 °C
- :1,34 g/cm³ (20 °C)
- :Omgivande
- :200-315-5
- :01-2119463277-33
- Förpackningstyp:Glass bottle for solids
- Förp.:250 g