Air Standard 1 * 1 Items
Katalog # REST34475
Air Standard 1 * 1 Items
Discontinued
Product Details & Documents
Metanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade för HPLC
#: 20864.320
Leverantör: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
Leverantör: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
#: 1.00030.2500
Leverantör: Merck
Natriumhydroxid 0.1 mol/l (0.1 N) i vattenlösning, AVS TITRINORM Reag. Ph. Eur. volumetrisk lösning
#: 31770.294
Leverantör: VWR Chemicals
Vatten, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
Leverantör: VWR Chemicals
Buffertlösningar, klara att användas, referenstemperatur 20 °C, Certipur®, Supelco®
#: 1.09439.1000
Leverantör: Merck



