Proppar, rombisk form
45/40
Catalog # 217-0809
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Hexagonal
- Flat
- Diamond-ground
Specifications
- NS:45/40
- Seq:13
DURAN®, borosilicate glass 3.3, clear.