Orders LinkContactUsLinkComponent
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 250 mm, 18/22 mm
Katalog # 231-0300
Leverantör:   RSG
Pharmacist's spatulas
Katalog # 231-0300
Leverantör:  RSG
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 250 mm, 18/22 mm
Price
530,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 2 Items
Product Details & Documents
Stainless steel blade, wooden handle.
  • Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
  • 75 to 150 mm: conical blade
Specifikationer
  • Beskrivning:
    Hardened blade
  • Bladbredd:
    18/22 mm
  • Klinge, längd:
    250 mm