Dropptratt
25 ml, 14/23 NS, 14/23 NS, 1 ml
Katalog # LENZ04172211
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
DURAN®, borosilicate glass, clear, with PTFE key.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Cylindrical shape. With standard ground joint cone and socket. Clearly visible white divisions for approximate volume reading.
DIN ISO 4800
Certifications: DIN ISO 4800
Documents
BSE TSE Glass Columns
BSE/TSE-certifikat
Specifikationer
- Kärna:14/23 NS
- NS-hylsa:14/23 NS
- Kapacitet:25 ml
- Indelning:1 ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



