Orders LinkContactUsLinkComponent
Empty solid phase extraction columns, CHROMABOND®
Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns, 70 ml
  MANA730382
Empty solid phase extraction columns, CHROMABOND®
  MANA730382
Empty solid phase extraction columns, CHROMABOND®
Empty solid phase extraction columns, CHROMABOND®
Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns, 70 ml
1 980,00 kr
1 * 10 Items
Empty and reservoir columns with accessories for solid phase extraction.
  • Empty PP and glass columns for individual packing with CHROMABOND® or other adsorbents
  • Reservoir columns from PP for application of medium-size samples
  • Accessories for SPE columns (filter elements, adapters)
  • Kapacitet:
    70 ml
  • Beskrivning:
    Reservoir columns, PP, with one adapter for 1, 3, 6 ml CHROMABOND® PP columns
  • Seq:
    16
Accessories
  •  
    Beskrivning
  • MANA732019
    PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight
    1 * 250 Items - 2 160,00 kr
  •  
    Beskrivning
  • MANA732020
    PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight
    1 * 250 Items - 3 030,00 kr
  •  
    Beskrivning
  • MANA730101
    Adapters (PP) for PP columns (1, 3 and 6 ml)
    1 * 10 Items - 1 580,00 kr
  •  
    Beskrivning
  • MANA730105
    Adapters (PVDF) for glass columns (3 and 6 ml)
    1 * 10 Items - 3 270,00 kr
  •  
    Beskrivning
  • MANA730026
    PE frits for PP columns, 70 ml
    1 * 250 Items - 4 600,00 kr
  •  
    Beskrivning
  • MANA730192
    Glass fiber frits for glass columns, 6 ml
    1 * 250 Items - 3 900,00 kr
  •  
    Beskrivning
  • MANA730351
    PE frits for PP columns, 15 ml
    1 * 250 Items - 3 900,00 kr
  •  
    Beskrivning
  • MANA730356
    PE frits for PP columns, 45 ml
    1 * 250 Items - 4 470,00 kr
  •  
    Beskrivning
  • MANA730475
    PE frits for PP columns, 150 ml
    1 * 250 Items - 4 940,00 kr
  •  
    Beskrivning
  • MANA730034
    PE frits for PP columns, 30 ml
    1 * 250 Items - 4 220,00 kr
  •  
    Beskrivning
  • MANA730566
    Adapters (PE) for PP columns (30 and 70 ml)
    1 * 1 KIT - 476,00 kr
  •  
    Beskrivning
  • MANA730385
    Adapters (PE) for PP columns (15, 45 and 70 ml)
    1 * 10 Items - 2 050,00 kr
  •  
    Beskrivning
  • MANA730191
    Glass fiber frits for glass columns, 3 ml
    1 * 250 Items - 3 210,00 kr
  •  
    Beskrivning
  • MANA730164
    PE frits for PP columns, 1 ml
    1 * 250 Items - 2 130,00 kr
  •  
    Beskrivning
  • MANA730163
    PE frits for PP columns, 6 ml
    1 * 250 Items - 3 700,00 kr
  •  
    Beskrivning
  • MANA730162
    PE frits for PP columns, 3 ml
    1 * 250 Items - 3 010,00 kr