Reference electrodes, ScienceLine
Enkel referenselektrod, ScienceLine, Ag/AgCl-referenssystem, B 2920+ Ag/AgCl, 0 - 14, −5...+100 °C, Glas, KCl 3 mol/l, Platina, 12×120 mm
Katalog # SCOI1070046
Leverantör: SI ANALYTICS
Gå till:
Product Details & Documents
Reference electrodes with glass shaft.
Specifikationer
- Ø×L:12×120 mm
- Typ:B 2920+ Ag/AgCl
- Beskrivning:Enkel referenselektrod, ScienceLine, Ag/AgCl-referenssystem
- Material:Glas
- Diafragma:Platina
- Elektrolyt:KCl 3 mol/l
- Temp:−5...+100 °C
- pH:0 - 14