Counting chambers
Counting chamber, dark lined, Nageotte, 0,5 mm
Katalog # 630-1511
Leverantör: MARIENFELD
Gå till:
Produktdetaljer och dokument
These counting chambers are suitable for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
- Different types of design available
Certifications: IVD 98/79/EC.
Leveransinformation: Supplied with two cover glasses of 0,4 mm thickness.
Dokument
Certificates
Sök efter LOT-nummerSpecifikationer
- Depth of chamber:0,5 mm
- Typ:Nageotte
- Beskrivning:Counting chamber, dark lined