Droppflaskor, WHEATON®
8-425, Gul
Catalog # 215-4040
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Gänga:8-425
- Seq:13
- Lock, färg:Gul