Orders LinkContactUsLinkComponent
Droppflaskor, WHEATON®
Droppflaskor, WHEATON®
8-425, Blå
Katalog # 215-4039
Leverantör:   DWK Life Sciences
Droppflaskor, WHEATON®
Katalog # 215-4039
Leverantör:  DWK Life Sciences
Droppflaskor, WHEATON®
Droppflaskor, WHEATON®
8-425, Blå
Price
5 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1.000 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifikationer
  • Gänga:
    8-425
  • Seq:
    12
  • Lock, färg:
    Blå