Trypsinizing flasks, WHEATON®
700 ml, Toppskruvlock 51-400, Skruvlock 20-415
Katalog # WHEA355757
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
Specifikationer
- Kapacitet:700 ml
- Beskrivning:Toppskruvlock 51-400
- Hals:Skruvlock 20-415