Droppflaskor, WHEATON®
20-410, Grå
Katalog # 215-4023
Leverantör: DWK Life Sciences
Gå till:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gänga:20-410
- Seq:41
- Lock, färg:Grå