Droppflaskor, WHEATON®
13-425, Blå
Katalog # 215-4031
Leverantör: DWK Life Sciences
Gå till:
Produktdetaljer och dokument
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifikationer
- Gänga:13-425
- Seq:18
- Lock, färg:Blå