Orders LinkContactUsLinkComponent
Adapters, bends
Adapters, bends
24/29 NS
Katalog # 201-0571
Leverantör:   Lenz Laborglas
Adapters, bends
Katalog # 201-0571
Leverantör:  Lenz Laborglas
Adapters, bends
Adapters, bends
24/29 NS
Price
630,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Inclined at an angle of 90°. With 2 standard ground cones.
Dokument
Specifikationer
  • Kärna:
    24/29 NS