Assortment Boxes, PS
Assortment box, with 18 compartments, Genomskinlig, 250×170×46 mm, PS
Katalog # HUEN613900
Leverantör: hünersdorff GmbH
Gå till:
Product Details & Documents
These assortment boxes made of high impact strength polystyrene (PS) are suitable for packaging purposes and create order in workshops and in household.
- Available either with versatile compartment subdivisions and versions or without compartments
- Transparent lid and bottom for optimum display of the contents or in colours suitable for foods
- Printing possible on request
Specifikationer
- Färg:Genomskinlig
- Beskrivning:Assortment box, with 18 compartments
- Material:PS
- Seq:11
- B×D×H:250×170×46 mm