Orders LinkContactUsLinkComponent
Trays
Trays
350×250×70 mm
Katalog # BOCH8736
Leverantör:   BOCHEM
Trays
Katalog # BOCH8736
Leverantör:  BOCHEM
Trays
Trays
350×250×70 mm
Price
1 200,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Tray made of stainless steel with two handles.
  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean
Multipurpose trays for a wide range of uses.
Specifikationer
  • Seq:
    02
  • B×D×H:
    350×250×70 mm