Trays
250×180×70 mm
Catalog # BOCH8735
Supplier: BOCHEM
Jump to:
Product Details & Documents
Tray made of stainless steel with two handles.
- Very good temperature resistance
- Very good chemical resistance
- Easy to clean
Multipurpose trays for a wide range of uses.
Specifications
- Seq:01
- B×D×H:250×180×70 mm