Sax
Trubbig, 155 mm
Katalog # BOCH4108
Leverantör: BOCHEM
Gå till:
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
- Mycket god temperaturtålighet, autoklaverbar
- God kemikalieresistens
- Tålig
Specifikationer
- Typ:Trubbig
- Längd:155 mm
- Seq:09