Sax
Spetsig, 145 mm
Katalog # 231-1293
Leverantör: BOCHEM
Gå till:
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
- Mycket god temperaturtålighet, autoklaverbar
- God kemikalieresistens
- Tålig
Specifikationer
- Typ:Spetsig
- Längd:145 mm
- Seq:05