Sax
Spetsig/trubbig, 145 mm
Catalog # 231-1291
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
- Mycket god temperaturtålighet, autoklaverbar
- God kemikalieresistens
- Tålig
Specifications
- Typ:Spetsig/trubbig
- Längd:145 mm
- Seq:02