Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
250 ml, 29/32 NS
Katalog # QVFLALG0050703
Leverantör:   QVF LABORTECHNIK
Separating funnels
Katalog # QVFLALG0050703
Leverantör:  QVF LABORTECHNIK
Separating funnels
250 ml, 29/32 NS
Price
1 850,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.
Specifikationer
  • NS-hylsa:
    29/32 NS
  • Kapacitet:
    250 ml
  • Seq:
    03