Orders LinkContactUsLinkComponent
ChromBox kit
ChromBox kit
Open end wrench, 6,35×7,93 mm
  JOURJR-804
ChromBox kit
  JOURJR-804
ChromBox kit
Open end wrench, 6,35×7,93 mm
390,00 kr
1 * 1 Items
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
Ordering information: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
  • Beskrivning:
    Open end wrench, 6,35×7,93 mm
  • Seq:
    05