Orders LinkContactUsLinkComponent
ChromBox kit
Deburring file
Katalog # JOURJR-802
Leverantör:   VICI JOUR RESEARCH
ChromBox kit
Katalog # JOURJR-802
Leverantör:  VICI JOUR RESEARCH
ChromBox kit
Deburring file
Price
505,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
 : Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifikationer
  • Beskrivning:
    Deburring file
  • Seq:
    04