Tubes and systems for active sampling
Diffusion samplers, ORSA 5
Katalog # DRAE6728891
Leverantör: Dräger
Gå till:
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Documents
Specifikationer
- Typ:ORSA 5
- Beskrivning:Diffusion samplers
- Seq:11