Order Entry
Sweden
Orders LinkContactUsLinkComponent
Kiselgel ≥99.0%

Kiselgel ≥99.0%

Plastic bottle for solids, 250 g
Catalog # 89709.30
Health
Kiselgel ≥99.0%
Catalog # 89709.30
CAS Number:  7631-86-9
Kiselgel ≥99.0%
Kiselgel ≥99.0%

Plastic bottle for solids, 250 g

CAS Number:  7631-86-9
Price
2 150,00 kr
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99.5% CAS No.: 7631-86-9 Molecular Formula: SiO2 Formula Weight: 60.09 Physical Form: -325 Mesh Fused Amorphous Powder, S.A. 5m2/g Density: 2.2 pH: 6 Melting Point: 1710° Boiling Point: 2230° Merck Reference: 14,8493 MDL No.: MFCD00011232

Synonyms:  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  • CAS Number:7631-86-9
  • Formula:SiO₂
  • MDL Number:MFCD00011232
  • Molecular Weight:60.08 g/mol
  • Merck Index:14,8493
  • Boiling Pt:2230 °C
  • Melting Pt:1710 °C
  • Density:2,2
  • Storage Temperature:Omgivande
  • EINECS:231-545-4
Specifications

Specifications

  • Förpackningstyp:
    Plastic bottle for solids
  • Förp.:
    250 g

Specifications