Scoops, Azlon®
Scoops, chemical resistand and food safe, 150 ml, 250 mm, HDPE
Katalog # AZLOSWL028
Leverantör: SciLabware
Gå till:
Produktdetaljer och dokument
Sturdy scoops made from natural rigid PE.
- Lightweight
- Good chemical resistance
- Food safe
Specifikationer
- Kapacitet:150 ml
- Beskrivning:Scoops, chemical resistand and food safe
- Längd:250 mm
- Material:HDPE
- Seq:01