Tilt Storage Boxes
9 lådor, Beige, 600×62×77 mm
Katalog # 216-4359
Leverantör: MULTIROIR
Gå till:
Product Details & Documents
ABS.
Each element contains a number of removable transparent drawers suitable for a wide range of items.
- Non toxic, suitable for use with foodstuffs
- Strong outer body high impact polystyrene plastic and inner compartments
- Can be fixed directly to the wall
Documents
VWR® For Your Space VWR® Lab bric-a-brac
Produktlitteratur
Specifikationer
- Färg:Beige
- Beskrivning:9 lådor
- Seq:08
- B×D×H:600×62×77 mm