Orders LinkContactUsLinkComponent
Tilt Storage Boxes
Tilt Storage Boxes
4 lådor, Beige, 600×168×207 mm
Catalog # 216-4356
Supplier:   MULTIROIR
Tilt Storage Boxes
Catalog # 216-4356
Supplier:  MULTIROIR
Tilt Storage Boxes
Tilt Storage Boxes
4 lådor, Beige, 600×168×207 mm
Price
1 300,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
ABS.
Each element contains a number of removable transparent drawers suitable for a wide range of items.
  • Non toxic, suitable for use with foodstuffs
  • Strong outer body high impact polystyrene plastic and inner compartments
  • Can be fixed directly to the wall
Documents
Specifications
  • Färg:
    Beige
  • Beskrivning:
    4 lådor
  • Seq:
    05
  • B×D×H:
    600×168×207 mm