Orders LinkContactUsLinkComponent
Tilt Storage Boxes
Tilt Storage Boxes
3 lådor, Beige, 600×197×240 mm
Katalog # 216-4348
Leverantör:   MULTIROIR
Tilt Storage Boxes
Katalog # 216-4348
Leverantör:  MULTIROIR
Tilt Storage Boxes
Tilt Storage Boxes
3 lådor, Beige, 600×197×240 mm
Price
1 575,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Product Details & Documents
ABS.
Each element contains a number of removable transparent drawers suitable for a wide range of items.
  • Non toxic, suitable for use with foodstuffs
  • Strong outer body high impact polystyrene plastic and inner compartments
  • Can be fixed directly to the wall
Documents
Specifikationer
  • Färg:
    Beige
  • Beskrivning:
    3 lådor
  • Seq:
    04
  • B×D×H:
    600×197×240 mm