Tilt Storage Boxes
2 lådor, Beige, 600×299×353 mm
Katalog # 216-4355
Leverantör: MULTIROIR
Gå till:
Product Details & Documents
ABS.
Each element contains a number of removable transparent drawers suitable for a wide range of items.
- Non toxic, suitable for use with foodstuffs
- Strong outer body high impact polystyrene plastic and inner compartments
- Can be fixed directly to the wall
Documents
VWR® For Your Space VWR® Lab bric-a-brac
Product Literature
Specifikationer
- Färg:Beige
- Beskrivning:2 lådor
- Seq:03
- B×D×H:600×299×353 mm