Measuring beaker with handle
1000 ml
Katalog # 213-0267
Leverantör: NICKEL ELECTRO
Gå till:
Product Details & Documents
Rostfritt stål, rostskyddad.
- Very good chemical resistance
- Very good thermal resistance
- Smooth surfaces for fast, easy cleaning
Conical, without rim, with broad base and textured graduations.
Specifikationer
- Kapacitet:1000 ml