Scoops
1000 ml, 340 mm
Katalog # 231-0352
Leverantör: NICKEL ELECTRO
Gå till:
Produktdetaljer och dokument
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
- Excellent thermal resistance, autoclavable
- Non magnetic, bright finish
- Highly polished surface makes cleaning easier
Specifikationer
- Kapacitet:1000 ml
- Längd:340 mm