Scoops
100 ml, 190 mm
Katalog # 231-0350
Leverantör: NICKEL ELECTRO
Gå till:
Produktdetaljer och dokument
Stainless steel scoops for collecting or measuring powders, pastes and granulates.
- Excellent thermal resistance, autoclavable
- Non magnetic, bright finish
- Highly polished surface makes cleaning easier
Specifikationer
- Kapacitet:100 ml
- Längd:190 mm