Orders LinkContactUsLinkComponent
Retort rods, without thread
Retort rods, without thread
12 mm, 1500 mm
Catalog # BOCH5125
Supplier:   BOCHEM
Retort rods, without thread
Catalog # BOCH5125
Supplier:  BOCHEM
Retort rods, without thread
Retort rods, without thread
12 mm, 1500 mm
Price
620,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • Rods for stand bases
  • Without thread
Specifications
  • Ø:
    12 mm
  • Längd:
    1500 mm
  • Seq:
    07