Cooling systems, CoolSafe™
CoolSafe™ box, PS, 140×216×165 mm
Katalog # DIVECSF-BOX
Leverantör: DIVERSIFIED BIOTECH
Gå till:
Product Details & Documents
Polystyrene cool safe box, aluminium cooling chamber.
- Durable
- Temperature stable
- Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.
Specifikationer
- Beskrivning:CoolSafe™ box, PS
- Mått:140×216×165 mm
- Seq:01