Orders LinkContactUsLinkComponent
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™
Cool brick for 1 °C, 5 ml tubes
Catalog # DIVEBRIK-1501
Supplier:   DIVERSIFIED BIOTECH
Cooling systems, CoolSafe™
Catalog # DIVEBRIK-1501
Cooling systems, CoolSafe™
Cooling systems, CoolSafe™
Cool brick for 1 °C, 5 ml tubes
Price
995,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Polystyrene cool safe box, aluminium cooling chamber.
  • Durable
  • Temperature stable
  • Corrosion resistant
Cooling system designed for 0,2 ml tubes (strips), 96 and 384 well microplates, 0,5, 1,5 and 5,0 ml tubes. Cool bricks with different temperature zones 1 or −10 °C.
Specifications
  • För:
    5 ml tubes
  • Beskrivning:
    Cool brick for 1 °C
  • Seq:
    14