Seizing forceps
Tumour and tissue forceps, No. 1, with loops (inner Ø 4,8 mm; outer Ø 6 mm), Straight, 90 mm
Katalog # HAMMHSC701-96
Leverantör: HAMMACHER KARL
Gå till:
Product Details & Documents
Forceps for taking hold of tumour and tissue.
Specifikationer
- Typ:Straight
- Beskrivning:Tumour and tissue forceps, No. 1, with loops (inner Ø 4,8 mm; outer Ø 6 mm)
- Längd:90 mm
- Seq:09