Seizing forceps
Tumour and tissue forceps, No. 2, with loops (inner Ø 2,2 mm; outer Ø 3 mm), Straight, 90 mm
Catalog # HAMMHSC700-93
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Forceps for taking hold of tumour and tissue.
Specifications
- Typ:Straight
- Beskrivning:Tumour and tissue forceps, No. 2, with loops (inner Ø 2,2 mm; outer Ø 3 mm)
- Längd:90 mm
- Seq:08